Assay Details
Gene Symbol: LOC23687777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854883.1 | XP_021710575.1 | 4 - 5 | 6914 | 61 | |
XM_021854884.1 | XP_021710576.1 | 5 - 6 | 6717 | 61 | ||
XM_021854885.1 | XP_021710577.1 | 5 - 6 | 6722 | 61 | ||
XM_021854886.1 | XP_021710578.1 | 4 - 5 | 6103 | 61 | ||
XM_021854887.1 | XP_021710579.1 | 4 - 5 | 6168 | 61 |

More Information
Set Membership: |
![]() ![]() |
