Assay Details
Gene Symbol: LOC23687777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854883.1 | XP_021710575.1 | 5 - 6 | 7276 | 88 | |
XM_021854884.1 | XP_021710576.1 | 6 - 7 | 7079 | 88 | ||
XM_021854885.1 | XP_021710577.1 | 6 - 7 | 7084 | 88 | ||
XM_021854886.1 | XP_021710578.1 | 5 - 6 | 6465 | 88 | ||
XM_021854887.1 | XP_021710579.1 | 5 - 6 | 6530 | 88 |
