Assay Details
Gene Symbol: LOC5564786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649077.2 | XP_001649127.1 | 6 - 7 | 1148 | 74 | |
XM_021851923.1 | XP_021707615.1 | 6 - 7 | 1373 | 74 | ||
XM_021851924.1 | XP_021707616.1 | 6 - 7 | 1187 | 74 |
