Assay Details
Gene Symbol: LOC5569239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652750.2 | XP_001652800.2 | 2 - 3 | 625 | 81 | |
XM_001652751.2 | XP_001652801.2 | 1 - 2 | 120 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
