Assay Details
Gene Symbol: LOC5568617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494738.2 | XP_011493040.1 | 2 - 3 | 304 | 84 | |
XM_021847059.1 | XP_021702751.1 | 2 - 3 | 280 | 84 | ||
XM_021847060.1 | XP_021702752.1 | 2 - 3 | 211 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011494738.2, XM_021847059.1, XM_021847060.1 transcripts.
Set Membership: |
![]() ![]() |
