Assay Details
Gene Symbol: LOC5568617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652399.2 | XP_001652449.1 | 7 - 8 | 1242 | 84 | |
XM_011494738.2 | XP_011493040.1 | 7 - 8 | 1164 | 84 | ||
XM_021847059.1 | XP_021702751.1 | 7 - 8 | 1140 | 84 | ||
XM_021847060.1 | XP_021702752.1 | 7 - 8 | 1071 | 84 | ||
XM_021847061.1 | XP_021702753.1 | 6 - 7 | 886 | 84 | ||
XM_021847062.1 | XP_021702754.1 | 6 - 7 | 949 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
