Assay Details
Gene Symbol: LOC5580331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844809.1 | XP_021700501.1 | 7 - 8 | 3253 | 77 | |
XM_021844810.1 | XP_021700502.1 | 5 - 6 | 1393 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
