Assay Details
Gene Symbol: LOC5573900 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847642.1 | XP_021703334.1 | 4 - 5 | 827 | 74 | |
XM_021847643.1 | XP_021703335.1 | 4 - 5 | 621 | 74 | ||
XM_021847644.1 | XP_021703336.1 | 4 - 5 | 555 | 74 | ||
XM_021847645.1 | XP_021703337.1 | 3 - 4 | 604 | 74 | ||
XM_021847646.1 | XP_021703338.1 | 4 - 5 | 516 | 74 | ||
XM_021847647.1 | XP_021703339.1 | 4 - 5 | 360 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
