Assay Details
Gene Symbol: LOC5573900 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847642.1 | XP_021703334.1 | 2 - 3 | 553 | 84 | |
XM_021847643.1 | XP_021703335.1 | 2 - 3 | 347 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
