Assay Details
Gene Symbol: LOC5571056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659448.2 | XP_001659498.2 | 11 - 12 | 2201 | 73 | |
XM_021852816.1 | XP_021708508.1 | 11 - 12 | 2233 | 73 | ||
XM_021852817.1 | XP_021708509.1 | 11 - 12 | 1974 | 73 | ||
XM_021852818.1 | XP_021708510.1 | 11 - 12 | 1986 | 73 | ||
XM_021852819.1 | XP_021708511.1 | 11 - 12 | 1871 | 73 | ||
XM_021852820.1 | XP_021708512.1 | 11 - 12 | 1908 | 73 | ||
XM_021852821.1 | XP_021708513.1 | 11 - 12 | 1945 | 73 | ||
XM_021852822.1 | XP_021708514.1 | 11 - 12 | 1854 | 73 | ||
XM_021852823.1 | XP_021708515.1 | 11 - 12 | 1895 | 73 | ||
XM_021852825.1 | XP_021708517.1 | 11 - 12 | 2220 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
