Assay Details
Gene Symbol: LOC5566966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651332.2 | XP_001651382.2 | 1 - 2 | 366 | 96 | |
XM_001651333.2 | XP_001651383.2 | 1 - 2 | 365 | 96 | ||
XM_001651334.2 | XP_001651384.1 | 1 - 2 | 365 | 96 | ||
XM_021845409.1 | XP_021701101.1 | 1 - 2 | 365 | 96 | ||
XM_021845410.1 | XP_021701102.1 | 1 - 2 | 365 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
