Assay Details
Gene Symbol: LOC5574842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661622.2 | XP_001661672.2 | 2 - 3 | 320 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5574842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661622.2 | XP_001661672.2 | 2 - 3 | 320 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |