Assay Details
Gene Symbol: LOC5565269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649543.2 | XP_001649593.2 | 1 - 2 | 539 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5565269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649543.2 | XP_001649593.2 | 1 - 2 | 539 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() |