Assay Details
Gene Symbol: LOC5574012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852676.1 | XP_021708368.1 | 1 - 2 | 857 | 71 | |
XM_021852720.1 | XP_021708412.1 | 1 - 2 | 862 | 71 | ||
XM_021852783.1 | XP_021708475.1 | 1 - 2 | 862 | 71 | ||
XM_021852839.1 | XP_021708531.1 | 1 - 2 | 858 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021852676.1, XM_021852720.1, XM_021852783.1, XM_021852839.1 transcripts.
Set Membership: |
![]() ![]() |
