Assay Details
Gene Symbol: LOC5572794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654145.2 | XP_001654195.2 | 2 - 3 | 595 | 60 | |
XM_021846989.1 | XP_021702681.1 | 2 - 3 | 592 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
