Assay Details
Gene Symbol: LOC5568747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658077.2 | XP_001658127.2 | 7 - 8 | 2175 | 96 | |
XM_021842362.1 | XP_021698054.1 | 7 - 8 | 2193 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
