Assay Details
Gene Symbol: LOC5566445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650766.2 | XP_001650816.1 | 3 - 4 | 1039 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5566445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650766.2 | XP_001650816.1 | 3 - 4 | 1039 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() |