Assay Details
Gene Symbol: LOC5575771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849104.1 | XP_021704796.1 | 1 - 2 | 154 | 81 | |
XM_021849107.1 | XP_021704799.1 | 1 - 2 | 154 | 81 | ||
XM_021849108.1 | XP_021704800.1 | 1 - 2 | 154 | 81 | ||
XM_021849109.1 | XP_021704801.1 | 1 - 2 | 154 | 81 | ||
XM_021849110.1 | XP_021704802.1 | 1 - 2 | 154 | 81 | ||
XM_021849111.1 | XP_021704803.1 | 1 - 2 | 154 | 81 | ||
XM_021849112.1 | XP_021704804.1 | 1 - 2 | 154 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021849104.1, XM_021849107.1, XM_021849108.1, XM_021849109.1, XM_021849110.1, XM_021849111.1, XM_021849112.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
