Assay Details
Gene Symbol: LOC5575771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849104.1 | XP_021704796.1 | 6 - 7 | 1116 | 59 | |
XM_021849105.1 | XP_021704797.1 | 6 - 7 | 1038 | 59 | ||
XM_021849107.1 | XP_021704799.1 | 6 - 7 | 1116 | 59 | ||
XM_021849108.1 | XP_021704800.1 | 5 - 6 | 1104 | 59 | ||
XM_021849109.1 | XP_021704801.1 | 6 - 7 | 1116 | 59 | ||
XM_021849110.1 | XP_021704802.1 | 5 - 6 | 1104 | 59 | ||
XM_021849111.1 | XP_021704803.1 | 5 - 6 | 1104 | 59 | ||
XM_021849112.1 | XP_021704804.1 | 6 - 7 | 1116 | 59 | ||
XM_021849113.1 | XP_021704805.1 | 3 - 4 | 668 | 59 | ||
XM_021849114.1 | XP_021704806.1 | 4 - 5 | 680 | 59 | ||
XM_021849115.1 | XP_021704807.1 | 3 - 4 | 576 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
