Assay Details
Gene Symbol: LOC5575714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662111.2 | XP_001662161.1 | 2 - 3 | 328 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5575714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662111.2 | XP_001662161.1 | 2 - 3 | 328 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() |