Assay Details
Gene Symbol: LOC5574541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655144.2 | XP_001655194.1 | 4 - 5 | 886 | 79 | |
XM_021843576.1 | XP_021699268.1 | 4 - 5 | 892 | 79 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5574541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655144.2 | XP_001655194.1 | 4 - 5 | 886 | 79 | |
XM_021843576.1 | XP_021699268.1 | 4 - 5 | 892 | 79 |