Assay Details
Gene Symbol: LOC5574541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655144.2 | XP_001655194.1 | 1 - 2 | 380 | 71 | |
XM_021843576.1 | XP_021699268.1 | 1 - 2 | 386 | 71 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5574541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655144.2 | XP_001655194.1 | 1 - 2 | 380 | 71 | |
XM_021843576.1 | XP_021699268.1 | 1 - 2 | 386 | 71 |