Assay Details
Gene Symbol: LOC5575906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844421.1 | XP_021700113.1 | 9 - 10 | 2031 | 100 | |
XM_021844422.1 | XP_021700114.1 | 9 - 10 | 2031 | 100 | ||
XM_021844423.1 | XP_021700115.1 | 9 - 10 | 2028 | 100 | ||
XM_021844424.1 | XP_021700116.1 | 8 - 9 | 1934 | 100 | ||
XM_021844425.1 | XP_021700117.1 | 8 - 9 | 1931 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
