Assay Details
Gene Symbol: LOC5575906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844421.1 | XP_021700113.1 | 6 - 7 | 1351 | 75 | |
XM_021844422.1 | XP_021700114.1 | 6 - 7 | 1351 | 75 | ||
XM_021844423.1 | XP_021700115.1 | 6 - 7 | 1348 | 75 | ||
XM_021844424.1 | XP_021700116.1 | 6 - 7 | 1350 | 75 | ||
XM_021844425.1 | XP_021700117.1 | 6 - 7 | 1347 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
