Assay Details
Gene Symbol: LOC5572482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660380.2 | XP_001660430.2 | 7 - 8 | 1580 | 71 | |
XM_021850427.1 | XP_021706119.1 | 8 - 9 | 1588 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
