Assay Details
Gene Symbol: LOC5572482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660380.2 | XP_001660430.2 | 3 - 4 | 890 | 102 | |
XM_021850427.1 | XP_021706119.1 | 4 - 5 | 898 | 102 |

More Information
Set Membership: |
![]() ![]() |
