Assay Details
Gene Symbol: LOC5574516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852475.1 | XP_021708167.1 | 1 - 2 | 204 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021852475.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
