Assay Details
Gene Symbol: LOC5570794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653246.2 | XP_001653296.1 | 2 - 3 | 645 | 108 | |
XM_021842910.1 | XP_021698602.1 | 2 - 3 | 625 | 108 | ||
XM_021842911.1 | XP_021698603.1 | 2 - 3 | 653 | 108 |

More Information
Set Membership: |
![]() ![]() |
