Assay Details
Gene Symbol: LOC5570794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653246.2 | XP_001653296.1 | 3 - 4 | 830 | 51 | |
XM_021842910.1 | XP_021698602.1 | 3 - 4 | 810 | 51 | ||
XM_021842911.1 | XP_021698603.1 | 3 - 4 | 838 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
