Assay Details
Gene Symbol: LOC5572780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654128.2 | XP_001654178.2 | 2 - 3 | 371 | 76 | |
XM_021852461.1 | XP_021708153.1 | 2 - 3 | 371 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
