Assay Details
Gene Symbol: LOC5575737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840026.1 | XP_021695718.1 | 10 - 11 | 3301 | 91 | |
XM_021840027.1 | XP_021695719.1 | 11 - 12 | 3208 | 91 | ||
XM_021840029.1 | XP_021695721.1 | 11 - 12 | 2836 | 91 | ||
XM_021840030.1 | XP_021695722.1 | 9 - 10 | 2881 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
