Assay Details
Gene Symbol: LOC5575737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840026.1 | XP_021695718.1 | 1 - 2 | 624 | 80 | |
XM_021840027.1 | XP_021695719.1 | 2 - 3 | 531 | 80 | ||
XM_021840030.1 | XP_021695722.1 | 1 - 2 | 624 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021840026.1, XM_021840027.1, XM_021840030.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
