Assay Details
Gene Symbol: LOC5575737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840026.1 | XP_021695718.1 | 2 - 3 | 1599 | 63 | |
XM_021840027.1 | XP_021695719.1 | 3 - 4 | 1506 | 63 | ||
XM_021840029.1 | XP_021695721.1 | 3 - 4 | 1134 | 63 | ||
XM_021840030.1 | XP_021695722.1 | 2 - 3 | 1599 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
