Assay Details
Gene Symbol: LOC5575737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840026.1 | XP_021695718.1 | 5 - 6 | 2246 | 78 | |
XM_021840027.1 | XP_021695719.1 | 6 - 7 | 2153 | 78 | ||
XM_021840029.1 | XP_021695721.1 | 6 - 7 | 1781 | 78 | ||
XM_021840030.1 | XP_021695722.1 | 5 - 6 | 2246 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
