Assay Details
Gene Symbol: LOC5575737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840026.1 | XP_021695718.1 | 6 - 7 | 2349 | 62 | |
XM_021840027.1 | XP_021695719.1 | 7 - 8 | 2256 | 62 | ||
XM_021840029.1 | XP_021695721.1 | 7 - 8 | 1884 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
