Assay Details
Gene Symbol: LOC5575737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840026.1 | XP_021695718.1 | 7 - 8 | 2772 | 80 | |
XM_021840027.1 | XP_021695719.1 | 8 - 9 | 2679 | 80 | ||
XM_021840029.1 | XP_021695721.1 | 8 - 9 | 2307 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
