Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 14 - 15 | 4237 | 62 | |
XM_021845538.1 | XP_021701230.1 | 14 - 15 | 4192 | 62 | ||
XM_021845539.1 | XP_021701231.1 | 13 - 14 | 3966 | 62 | ||
XM_021845540.1 | XP_021701232.1 | 13 - 14 | 3037 | 62 | ||
XM_021845541.1 | XP_021701233.1 | 13 - 14 | 2992 | 62 | ||
XM_021845543.1 | XP_021701235.1 | 12 - 13 | 2701 | 62 | ||
XM_021845544.1 | XP_021701236.1 | 12 - 13 | 2656 | 62 | ||
XM_021845545.1 | XP_021701237.1 | 3 - 4 | 390 | 62 |

More Information
Set Membership: |
![]() ![]() |
