Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 19 - 20 | 6386 | 73 | |
XM_021845538.1 | XP_021701230.1 | 19 - 20 | 6341 | 73 | ||
XM_021845539.1 | XP_021701231.1 | 18 - 19 | 6115 | 73 | ||
XM_021845540.1 | XP_021701232.1 | 18 - 19 | 5186 | 73 | ||
XM_021845541.1 | XP_021701233.1 | 18 - 19 | 5141 | 73 | ||
XM_021845542.1 | XP_021701234.1 | 18 - 19 | 5033 | 73 | ||
XM_021845543.1 | XP_021701235.1 | 17 - 18 | 4850 | 73 | ||
XM_021845544.1 | XP_021701236.1 | 17 - 18 | 4805 | 73 | ||
XM_021845545.1 | XP_021701237.1 | 8 - 9 | 2539 | 73 |
