Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 20 - 21 | 6495 | 61 | |
XM_021845538.1 | XP_021701230.1 | 20 - 21 | 6450 | 61 | ||
XM_021845539.1 | XP_021701231.1 | 19 - 20 | 6224 | 61 | ||
XM_021845540.1 | XP_021701232.1 | 19 - 20 | 5295 | 61 | ||
XM_021845541.1 | XP_021701233.1 | 19 - 20 | 5250 | 61 | ||
XM_021845542.1 | XP_021701234.1 | 19 - 20 | 5142 | 61 | ||
XM_021845543.1 | XP_021701235.1 | 18 - 19 | 4959 | 61 | ||
XM_021845544.1 | XP_021701236.1 | 18 - 19 | 4914 | 61 | ||
XM_021845545.1 | XP_021701237.1 | 9 - 10 | 2648 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
