Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 8 - 9 | 2120 | 104 | |
XM_021845539.1 | XP_021701231.1 | 7 - 8 | 1849 | 104 | ||
XM_021845542.1 | XP_021701234.1 | 8 - 9 | 2120 | 104 |

More Information
Set Membership: |
![]() ![]() |
