Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 10 - 11 | 3659 | 67 | |
XM_021845538.1 | XP_021701230.1 | 10 - 11 | 3614 | 67 | ||
XM_021845539.1 | XP_021701231.1 | 9 - 10 | 3388 | 67 | ||
XM_021845540.1 | XP_021701232.1 | 9 - 10 | 2459 | 67 | ||
XM_021845541.1 | XP_021701233.1 | 9 - 10 | 2414 | 67 | ||
XM_021845542.1 | XP_021701234.1 | 10 - 11 | 3659 | 67 |

More Information
Set Membership: |
![]() ![]() |
