Assay Details
Gene Symbol: LOC5567089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841760.1 | XP_021697452.1 | 2 - 3 | 769 | 75 | |
XM_021841761.1 | XP_021697453.1 | 2 - 3 | 768 | 75 | ||
XM_021841762.1 | XP_021697454.1 | 2 - 3 | 642 | 75 | ||
XM_021841763.1 | XP_021697455.1 | 3 - 4 | 876 | 75 | ||
XM_021841764.1 | XP_021697456.1 | 3 - 4 | 856 | 75 | ||
XM_021841765.1 | XP_021697457.1 | 2 - 3 | 675 | 75 | ||
XM_021841766.1 | XP_021697458.1 | 2 - 3 | 649 | 75 | ||
XM_021841767.1 | XP_021697459.1 | 2 - 3 | 557 | 75 | ||
XM_021841768.1 | XP_021697460.1 | 2 - 3 | 555 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
