Assay Details
Gene Symbol: LOC5574228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841865.1 | XP_021697557.1 | 3 - 4 | 1514 | 79 | |
XM_021841866.1 | XP_021697558.1 | 3 - 4 | 1532 | 79 | ||
XM_021841868.1 | XP_021697560.1 | 3 - 4 | 1458 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
