Assay Details
Gene Symbol: LOC5571651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845509.1 | XP_021701201.1 | 12 - 13 | 5029 | 101 | |
XM_021845510.1 | XP_021701202.1 | 13 - 14 | 4926 | 101 | ||
XM_021845511.1 | XP_021701203.1 | 13 - 14 | 4736 | 101 | ||
XM_021845512.1 | XP_021701204.1 | 13 - 14 | 4831 | 101 | ||
XM_021845513.1 | XP_021701205.1 | 12 - 13 | 5004 | 101 | ||
XM_021845514.1 | XP_021701206.1 | 12 - 13 | 5029 | 101 | ||
XM_021845515.1 | XP_021701207.1 | 13 - 14 | 4963 | 101 | ||
XM_021845516.1 | XP_021701208.1 | 13 - 14 | 4937 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
