Assay Details
Gene Symbol: LOC5571651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845509.1 | XP_021701201.1 | 2 - 3 | 1182 | 82 | |
XM_021845510.1 | XP_021701202.1 | 3 - 4 | 1079 | 82 | ||
XM_021845511.1 | XP_021701203.1 | 3 - 4 | 889 | 82 | ||
XM_021845512.1 | XP_021701204.1 | 3 - 4 | 984 | 82 | ||
XM_021845513.1 | XP_021701205.1 | 2 - 3 | 1184 | 82 | ||
XM_021845514.1 | XP_021701206.1 | 2 - 3 | 1182 | 82 | ||
XM_021845515.1 | XP_021701207.1 | 2 - 3 | 1182 | 82 | ||
XM_021845516.1 | XP_021701208.1 | 2 - 3 | 1183 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
