Assay Details
Gene Symbol: LOC5571651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845509.1 | XP_021701201.1 | 4 - 5 | 1407 | 81 | |
XM_021845510.1 | XP_021701202.1 | 5 - 6 | 1304 | 81 | ||
XM_021845511.1 | XP_021701203.1 | 5 - 6 | 1114 | 81 | ||
XM_021845512.1 | XP_021701204.1 | 5 - 6 | 1209 | 81 | ||
XM_021845513.1 | XP_021701205.1 | 4 - 5 | 1409 | 81 | ||
XM_021845514.1 | XP_021701206.1 | 4 - 5 | 1407 | 81 | ||
XM_021845515.1 | XP_021701207.1 | 4 - 5 | 1407 | 81 | ||
XM_021845516.1 | XP_021701208.1 | 4 - 5 | 1408 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
