Assay Details
Gene Symbol: LOC5575110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661782.2 | XP_001661832.1 | 3 - 4 | 592 | 66 | |
XM_021851003.1 | XP_021706695.1 | 4 - 5 | 1293 | 66 | ||
XM_021851004.1 | XP_021706696.1 | 4 - 5 | 1293 | 66 |

More Information
Set Membership: |
![]() ![]() |
