Assay Details
Gene Symbol: LOC5571858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847232.1 | XP_021702924.1 | 2 - 3 | 650 | 88 | |
XM_021847233.1 | XP_021702925.1 | 4 - 5 | 2008 | 88 | ||
XM_021847234.1 | XP_021702926.1 | 3 - 4 | 1243 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021847232.1, XM_021847233.1, XM_021847234.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
