Assay Details
Gene Symbol: LOC5571858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847232.1 | XP_021702924.1 | 19 - 20 | 3377 | 67 | |
XM_021847233.1 | XP_021702925.1 | 21 - 22 | 4735 | 67 | ||
XM_021847234.1 | XP_021702926.1 | 20 - 21 | 3970 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
