Assay Details
Gene Symbol: LOC5571858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847232.1 | XP_021702924.1 | 17 - 18 | 2713 | 72 | |
XM_021847233.1 | XP_021702925.1 | 19 - 20 | 4071 | 72 | ||
XM_021847234.1 | XP_021702926.1 | 18 - 19 | 3306 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
